siRNA Details
| Gene Symbol: Plekha6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249816.2 | XP_006249878.1 | 2 | 3636 | ||
| XM_006249817.2 | XP_006249879.1 | 2 | 3634 | |||
| XM_006249818.2 | XP_006249880.1 | 2 | 3633 | |||
| XM_008769515.1 | XP_008767737.1 | 3 | 152 | |||
| XM_341118.7 | XP_341119.5 | 2 | 3636 | |||

Plekha6
Genomic Map