siRNA Details
| Gene Symbol: Zc3h11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047902.1 | NP_001041367.1 | 3 | 353 | ||
| XM_008769482.1 | XP_008767704.1 | 1 | 88 | |||
| XM_008769483.1 | XP_008767705.1 | 1 | 88 | |||
| XM_008769484.1 | XP_008767706.1 | 1 | 106 | |||
| GenBank mRNA | DQ480752.1 | - | 3 | 353 | ||

Zc3h11a
Genomic Map