siRNA Details
| Gene Symbol: Zc3h11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047902.1 | NP_001041367.1 | 12 | 1582 | ||
| XM_008769482.1 | XP_008767704.1 | 10 | 1359 | |||
| XM_008769483.1 | XP_008767705.1 | 10 | 1356 | |||
| XM_008769484.1 | XP_008767706.1 | 10 | 1332 | |||
| GenBank mRNA | BC111404.1 | - | 5 | 595 | ||
| DQ480752.1 | - | 12 | 1582 | |||

Zc3h11a
Genomic Map