siRNA Details
| Gene Symbol: Ube2t | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108344.2 | NP_001101814.2 | 6 | 610 | ||
| GenBank mRNA | BC167012.1 | - | 6 | 571 | 9099623 | |
| FQ233263.1 | - | 6 | 598 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2t| Gene Symbol: Ube2t | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108344.2 | NP_001101814.2 | 6 | 610 | ||
| GenBank mRNA | BC167012.1 | - | 6 | 571 | 9099623 | |
| FQ233263.1 | - | 6 | 598 | |||