siRNA Details
| Gene Symbol: RGD1566099 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108347.1 | NP_001101817.1 | 1 | 204 | ||
| GenBank mRNA | BC166874.1 | - | 1 | 201 | 6890641 | |
| BC166876.1 | - | 1 | 201 | 6919569 | ||
| BC166924.1 | - | 1 | 201 | 6919449 | ||

RGD1566099
Genomic Map