siRNA Details
| Gene Symbol: Arpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025717.1 | NP_001020888.1 | 2 | 219 | ||
| GenBank mRNA | BC099238.1 | - | 2 | 219 | 7445509 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arpc5| Gene Symbol: Arpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025717.1 | NP_001020888.1 | 2 | 219 | ||
| GenBank mRNA | BC099238.1 | - | 2 | 219 | 7445509 | |