siRNA Details
Gene Symbol: Arpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025717.1 | NP_001020888.1 | 3 | 375 | ||
GenBank mRNA | BC099238.1 | - | 3 | 375 | 7445509 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025717.1 | NP_001020888.1 | 3 | 375 | ||
GenBank mRNA | BC099238.1 | - | 3 | 375 | 7445509 |