siRNA Details
| Gene Symbol: Rfwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025126.1 | NP_001020297.1 | 5 | 607 | ||
| XM_006250113.2 | XP_006250175.1 | 12 | 1806 | |||
| XM_006250114.2 | XP_006250176.1 | 12 | 1794 | |||
| XM_006250115.2 | XP_006250177.1 | 11 | 1746 | |||
| XM_006250116.2 | XP_006250178.1 | 11 | 1734 | |||
| GenBank mRNA | BC091284.1 | - | 5 | 607 | 7304516 | |

Rfwd2
Genomic Map