siRNA Details
| Gene Symbol: GPAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003801.3 | NP_003792.1 | 5 | 657 | ||
| GenBank mRNA | AB002135.1 | - | 5 | 642 | ||
| AB006969.1 | - | 5 | 555 | |||
| AK312759.1 | - | 5 | 669 | |||
| AL157437.1 | - | 4 | 1154 | |||
| BC003171.1 | - | 5 | 633 | 2906111 | ||
| BC004129.2 | - | 5 | 657 | 3504366 | ||
| BC006383.2 | - | 4 | 430 | 4099139 | ||

GPAA1
Genomic Map