siRNA Details
| Gene Symbol: Sft2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034011.1 | NP_001029183.1 | 6, 7 | 424 | ||
| GenBank mRNA | BC099092.1 | - | 6, 7 | 424 | 7457938 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sft2d2| Gene Symbol: Sft2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034011.1 | NP_001029183.1 | 6, 7 | 424 | ||
| GenBank mRNA | BC099092.1 | - | 6, 7 | 424 | 7457938 | |