siRNA Details
| Gene Symbol: Mpzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007728.1 | NP_001007729.1 | 2 | 219 | ||
| XM_006250180.2 | XP_006250242.1 | 2 | 267 | |||
| GenBank mRNA | BC078916.1 | - | 2 | 219 | 7096438 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mpzl1| Gene Symbol: Mpzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007728.1 | NP_001007729.1 | 2 | 219 | ||
| XM_006250180.2 | XP_006250242.1 | 2 | 267 | |||
| GenBank mRNA | BC078916.1 | - | 2 | 219 | 7096438 | |