siRNA Details
| Gene Symbol: Klhdc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108350.1 | NP_001101820.1 | 3 | 1091 | ||
| XM_008769745.1 | XP_008767967.1 | 4 | 1724 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhdc9| Gene Symbol: Klhdc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108350.1 | NP_001101820.1 | 3 | 1091 | ||
| XM_008769745.1 | XP_008767967.1 | 4 | 1724 | |||