siRNA Details
| Gene Symbol: Adck3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013185.1 | NP_001013203.1 | 7, 8 | 985 | ||
| XM_006250387.1 | XP_006250449.1 | 7, 8 | 1068 | |||
| GenBank mRNA | BC091388.1 | - | 7, 8 | 985 | 7321446 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adck3| Gene Symbol: Adck3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013185.1 | NP_001013203.1 | 7, 8 | 985 | ||
| XM_006250387.1 | XP_006250449.1 | 7, 8 | 1068 | |||
| GenBank mRNA | BC091388.1 | - | 7, 8 | 985 | 7321446 | |