siRNA Details
| Gene Symbol: Enah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012150.1 | NP_001012150.1 | 9 | 1349 | ||
| XM_006250393.2 | XP_006250455.1 | 10 | 2417 | |||
| XM_006250400.2 | XP_006250462.1 | 10 | 1694 | |||
| XM_006250401.2 | XP_006250463.1 | 10 | 1694 | |||
| XM_008769859.1 | XP_008768081.1 | 12 | 2382 | |||
| XM_008769860.1 | XP_008768082.1 | 10 | 2943 | |||
| XM_008769861.1 | XP_008768083.1 | 10 | 2313 | |||
| GenBank mRNA | BC083927.1 | - | 9 | 1349 | 7135538 | |

Enah
Genomic Map