siRNA Details
| Gene Symbol: MYH13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003802.2 | NP_003793.2 | 23, 24 | 3032 | ||
| GenBank mRNA | AF111782.2 | - | 23, 24 | 3032 | ||
| AK294316.1 | - | 13, 14 | 1834 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYH13| Gene Symbol: MYH13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003802.2 | NP_003793.2 | 23, 24 | 3032 | ||
| GenBank mRNA | AF111782.2 | - | 23, 24 | 3032 | ||
| AK294316.1 | - | 13, 14 | 1834 | |||