siRNA Details
| Gene Symbol: MYOM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003803.3 | NP_003794.3 | 3 | 743 | ||
| NM_019856.1 | NP_062830.1 | 3 | 743 | |||
| XM_017026062.1 | XP_016881551.1 | 3 | 506 | |||
| GenBank mRNA | AJ621424.1 | - | 3 | 743 | ||
| AK125322.1 | - | 3 | 743 | |||
| AK302444.1 | - | 1 | 84 | |||
| BC116183.1 | - | 3 | 484 | 40073728 | ||
| X69090.1 | - | 1 | 118 | |||

MYOM1
Genomic Map