siRNA Details
| Gene Symbol: RGD1310587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100857.1 | NP_001094327.1 | 1 | 291 | ||
| GenBank mRNA | BC081867.1 | - | 1 | 291 | ||
| BC100157.1 | - | 1 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1310587| Gene Symbol: RGD1310587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100857.1 | NP_001094327.1 | 1 | 291 | ||
| GenBank mRNA | BC081867.1 | - | 1 | 291 | ||
| BC100157.1 | - | 1 | 291 | |||