siRNA Details
| Gene Symbol: MYOM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003803.3 | NP_003794.3 | 4 | 791 | ||
| NM_019856.1 | NP_062830.1 | 4 | 791 | |||
| XM_017026062.1 | XP_016881551.1 | 4 | 554 | |||
| GenBank mRNA | AJ621424.1 | - | 4 | 791 | ||
| AK125322.1 | - | 4 | 791 | |||
| AK302444.1 | - | 2 | 132 | |||
| BC116183.1 | - | 4 | 532 | 40073728 | ||
| X69090.1 | - | 2 | 166 | |||

MYOM1
Genomic Map