siRNA Details
| Gene Symbol: Sertad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108351.1 | NP_001101821.1 | 3 | 436 | ||
| XM_008769863.1 | XP_008768085.1 | 2 | 4342 | |||
| XM_008769864.1 | XP_008768086.1 | 3 | 3812 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sertad4| Gene Symbol: Sertad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108351.1 | NP_001101821.1 | 3 | 436 | ||
| XM_008769863.1 | XP_008768085.1 | 2 | 4342 | |||
| XM_008769864.1 | XP_008768086.1 | 3 | 3812 | |||