siRNA Details
| Gene Symbol: Fam69a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170456.1 | NP_001163927.1 | 2 | 195 | ||
| GenBank mRNA | AI706300.1 | - | 2 | 195 | ||
| BC166752.1 | - | 2 | 146 | 5621368 | ||
| FQ211286.1 | - | 384 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam69a| Gene Symbol: Fam69a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170456.1 | NP_001163927.1 | 2 | 195 | ||
| GenBank mRNA | AI706300.1 | - | 2 | 195 | ||
| BC166752.1 | - | 2 | 146 | 5621368 | ||
| FQ211286.1 | - | 384 | ||||