siRNA Details
| Gene Symbol: Helq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014134.2 | NP_001014156.2 | 2 | 657 | ||
| XM_006250666.2 | XP_006250728.1 | 2 | 654 | |||
| GenBank mRNA | BC086609.1 | - | 2 | 553 | ||
| BC158652.1 | - | 2 | 550 | 8368098 | ||

Helq
Genomic Map