siRNA Details
| Gene Symbol: Helq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014134.2 | NP_001014156.2 | 8 | 1854 | ||
| XM_006250666.2 | XP_006250728.1 | 8 | 1851 | |||
| XM_008770040.1 | XP_008768262.1 | 6 | 680 | |||
| XM_008770041.1 | XP_008768263.1 | 3 | 304 | |||

Helq
Genomic Map