siRNA Details
| Gene Symbol: Tmem150c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108354.2 | NP_001101824.1 | 6 | 337 | ||
| XM_006250670.2 | XP_006250732.1 | 6 | 672 | |||
| XM_006250671.2 | XP_006250733.1 | 6 | 2361 | |||
| GenBank mRNA | BC169066.1 | - | 6 | 337 | 5598025 | |
| FQ214560.1 | - | 6 | 354 | |||

Tmem150c
Genomic Map