siRNA Details
| Gene Symbol: Lnx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108358.1 | NP_001101828.1 | 9, 10 | 2079 | ||
| XM_006250902.2 | XP_006250964.1 | 9, 10 | 2073 | |||
| XM_006250903.2 | XP_006250965.1 | 8, 9 | 4111 | |||
| XM_006250904.2 | XP_006250966.1 | 8, 9 | 4081 | |||
| GenBank mRNA | EU219404.1 | - | 8, 9 | 1922 | ||
| EU219405.1 | - | 8, 9 | 1922 | |||
| EU219406.1 | - | 8, 9 | 1790 | |||

Lnx1
Genomic Map