siRNA Details
| Gene Symbol: Dcun1d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108359.1 | NP_001101829.1 | 4, 5 | 345 | ||
| XM_006250907.2 | XP_006250969.1 | 2, 3 | 244 | |||
| XM_008770158.1 | XP_008768380.1 | 4, 5 | 356 | |||
| XM_008770159.1 | XP_008768381.1 | 2, 3 | 147 | |||

Dcun1d4
Genomic Map