siRNA Details
| Gene Symbol: Tbc1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251004.2 | XP_006251066.1 | 3 | 1095 | ||
| XM_006251005.2 | XP_006251067.1 | 3 | 1095 | |||
| XM_006251006.2 | XP_006251068.1 | 2 | 489 | |||
| XM_006251007.1 | XP_006251069.1 | 2 | 513 | |||
| XM_341215.7 | XP_341216.4 | 3 | 1095 | |||

Tbc1d1
Genomic Map