siRNA Details
| Gene Symbol: Pcdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004087.1 | NP_001004087.1 | 1 | 319 | ||
| XM_006251017.2 | XP_006251079.1 | 1 | 397 | |||
| XM_006251022.2 | XP_006251084.1 | 1 | 397 | |||
| XM_006251023.2 | XP_006251085.1 | 1 | 397 | |||
| XM_008770189.1 | XP_008768411.1 | 1 | 397 | |||
| GenBank mRNA | AY690613.1 | - | 1 | 319 | ||
| AY690614.1 | - | 1 | 319 | |||
| AY690615.1 | - | 1 | 319 | |||
| AY690616.1 | - | 1 | 319 | |||

Pcdh7
Genomic Map