siRNA Details
| Gene Symbol: Wdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014135.1 | NP_001014157.1 | 4 | 347 | ||
| GenBank mRNA | AY986483.1 | - | 4 | 334 | ||
| BC085864.1 | - | 4 | 347 | 7190485 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr1| Gene Symbol: Wdr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014135.1 | NP_001014157.1 | 4 | 347 | ||
| GenBank mRNA | AY986483.1 | - | 4 | 334 | ||
| BC085864.1 | - | 4 | 347 | 7190485 | ||