siRNA Details
| Gene Symbol: Tmem128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108362.2 | NP_001101832.1 | 2 | 186 | ||
| XM_006251099.2 | XP_006251161.1 | 2 | 375 | |||
| GenBank mRNA | BC167007.1 | - | 2 | 186 | 7310387 | |
| FQ211361.1 | - | 2 | 198 | |||
| FQ213588.1 | - | 2 | 219 | |||
| FQ217844.1 | - | 2 | 213 | |||
| FQ233251.1 | - | 2 | 190 | |||

Tmem128
Genomic Map