siRNA Details
| Gene Symbol: Stx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012151.1 | NP_001012151.1 | 9 | 831 | ||
| GenBank mRNA | BC079183.1 | - | 9 | 831 | 7130966 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stx18| Gene Symbol: Stx18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012151.1 | NP_001012151.1 | 9 | 831 | ||
| GenBank mRNA | BC079183.1 | - | 9 | 831 | 7130966 | |