siRNA Details
| Gene Symbol: Hmx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108363.1 | NP_001101833.1 | 2 | 634 | ||
| XM_006251147.2 | XP_006251209.1 | 2 | 902 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmx1| Gene Symbol: Hmx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108363.1 | NP_001101833.1 | 2 | 634 | ||
| XM_006251147.2 | XP_006251209.1 | 2 | 902 | |||