siRNA Details
| Gene Symbol: TNFSF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003809.2 | NP_003800.1 | 7 | 1024 | ||
| NR_037146.1 | 8 | 1263 | ||||
| GenBank mRNA | AF030099.1 | - | 7 | 945 | ||
| AF055872.1 | - | 7 | 985 | |||
| AY358870.1 | - | 7 | 985 | |||
| BC019047.2 | - | 8 | 1263 | 4766071 | ||
| BC071837.1 | - | 8 | 1246 | 5934943 | ||
| BC104420.1 | - | 7 | 960 | 40004935 | ||

TNFSF12
Genomic Map