siRNA Details
| Gene Symbol: Cabp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007730.1 | NP_001007731.1 | 2, 3 | 689 | ||
| GenBank mRNA | AY841152.1 | - | 2, 3 | 302 | ||
| BC081959.1 | - | 2, 3 | 689 | 7124492 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cabp7| Gene Symbol: Cabp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007730.1 | NP_001007731.1 | 2, 3 | 689 | ||
| GenBank mRNA | AY841152.1 | - | 2, 3 | 302 | ||
| BC081959.1 | - | 2, 3 | 689 | 7124492 | ||