siRNA Details
| Gene Symbol: Ogdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017461.1 | NP_001017461.1 | 9 | 1199 | ||
| XM_006251446.2 | XP_006251508.1 | 9 | 1315 | |||
| XM_006251447.2 | XP_006251509.1 | 9 | 1461 | |||
| XM_006251448.2 | XP_006251510.1 | 9 | 1238 | |||
| GenBank mRNA | BC083811.1 | - | 9 | 1199 | 7110182 | |
| FQ222842.1 | - | 9 | 1174 | |||

Ogdh
Genomic Map