siRNA Details
| Gene Symbol: Tmem17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010961.1 | NP_001010961.1 | 2, 3 | 547 | ||
| GenBank mRNA | BC089059.1 | - | 2, 3 | 547 | 7320241 | |
| FQ228748.1 | - | 2, 3 | 205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem17| Gene Symbol: Tmem17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010961.1 | NP_001010961.1 | 2, 3 | 547 | ||
| GenBank mRNA | BC089059.1 | - | 2, 3 | 547 | 7320241 | |
| FQ228748.1 | - | 2, 3 | 205 | |||