siRNA Details
| Gene Symbol: Zswim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271275.1 | NP_001258204.1 | 1 | 378 | ||
| XM_008770546.1 | XP_008768768.1 | 1 | 390 | |||
| XM_008770547.1 | XP_008768769.1 | 1 | 390 | |||
| XM_008770548.1 | XP_008768770.1 | 1 | 389 | |||
| XM_008770549.1 | XP_008768771.1 | 1 | 390 | |||
| XM_008770550.1 | XP_008768772.1 | 1 | 390 | |||
| XM_008770551.1 | XP_008768773.1 | 1 | 390 | |||

Zswim8
Genomic Map