siRNA Details
| Gene Symbol: Chchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108369.1 | NP_001101839.1 | 3 | 310 | ||
| GenBank mRNA | BC168773.1 | - | 3 | 306 | 6920789 | |
| FQ217372.1 | - | 3 | 283 | |||
| FQ217729.1 | - | 3 | 290 | |||
| FQ221927.1 | - | 3 | 276 | |||
| FQ224582.1 | - | 3 | 291 | |||
| FQ224809.1 | - | 3 | 291 | |||

Chchd1
Genomic Map