siRNA Details
| Gene Symbol: Spetex-2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011702.1 | NP_001011702.1 | 141 | |||
| GenBank mRNA | AB180080.1 | - | 141 | |||
| Gene Symbol: Spetex-2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011698.1 | NP_001011698.1 | 2 | 141 | ||
| GenBank mRNA | AB180078.1 | - | 2 | 141 | ||
| Gene Symbol: Spetex-2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011701.1 | NP_001011701.1 | 2 | 141 | ||
| GenBank mRNA | AB180079.1 | - | 2 | 141 | ||

Spetex-2E
Genomic Map