siRNA Details
| Gene Symbol: Ngly1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014136.1 | NP_001014158.1 | 4 | 709 | ||
| GenBank mRNA | BC083837.1 | - | 4 | 709 | 7113173 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ngly1| Gene Symbol: Ngly1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014136.1 | NP_001014158.1 | 4 | 709 | ||
| GenBank mRNA | BC083837.1 | - | 4 | 709 | 7113173 | |