siRNA Details
| Gene Symbol: Atxn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251728.2 | XP_006251790.1 | 12 | 2751 | ||
| XM_008770600.1 | XP_008768822.1 | 12 | 2751 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atxn7| Gene Symbol: Atxn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251728.2 | XP_006251790.1 | 12 | 2751 | ||
| XM_008770600.1 | XP_008768822.1 | 12 | 2751 | |||