siRNA Details
Gene Symbol: Fam107a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025129.1 | NP_001020300.1 | 2 | 122 | ||
GenBank mRNA | BC093601.1 | - | 2 | 122 | 7316049 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam107a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025129.1 | NP_001020300.1 | 2 | 122 | ||
GenBank mRNA | BC093601.1 | - | 2 | 122 | 7316049 |