siRNA Details
| Gene Symbol: ASIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001672.2 | NP_001663.2 | 1 | 134 | ||
| XM_005260412.3 | XP_005260469.1 | 2 | 2190 | |||
| XM_011528820.2 | XP_011527122.1 | 3 | 313 | |||
| XM_011528821.1 | XP_011527123.1 | 3 | 271 | |||
| GenBank mRNA | BC104238.1 | - | 1 | 126 | 40033052 | |
| BC104239.2 | - | 1 | 126 | 40033056 | ||

ASIP
Genomic Map