siRNA Details
| Gene Symbol: RGD1563302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162899.1 | NP_001156371.1 | 2 | 459 | ||
| XM_006252122.2 | XP_006252184.1 | 2 | 513 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1563302| Gene Symbol: RGD1563302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162899.1 | NP_001156371.1 | 2 | 459 | ||
| XM_006252122.2 | XP_006252184.1 | 2 | 513 | |||