siRNA Details
| Gene Symbol: Cdadc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012156.1 | NP_001012156.1 | 1, 2 | 170 | ||
| XM_006252123.1 | XP_006252185.1 | 1, 2 | 218 | |||
| XM_006252125.2 | XP_006252187.1 | 1, 2 | 218 | |||
| XM_008770760.1 | XP_008768982.1 | 1, 2 | 220 | |||
| GenBank mRNA | BC083736.1 | - | 1, 2 | 170 | 7191631 | |
| FQ213068.1 | - | 1, 2 | 189 | |||

Cdadc1
Genomic Map