siRNA Details
| Gene Symbol: Ebpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108381.1 | NP_001101851.1 | 3, 4 | 391 | ||
| GenBank mRNA | FQ213140.1 | - | 3, 4 | 397 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ebpl| Gene Symbol: Ebpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108381.1 | NP_001101851.1 | 3, 4 | 391 | ||
| GenBank mRNA | FQ213140.1 | - | 3, 4 | 397 | ||