siRNA Details
| Gene Symbol: Entpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252275.2 | XP_006252337.1 | 7 | 969 | ||
| XM_006252276.2 | XP_006252338.1 | 7 | 1372 | |||
| XM_006252277.2 | XP_006252339.1 | 7 | 969 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Entpd4| Gene Symbol: Entpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252275.2 | XP_006252337.1 | 7 | 969 | ||
| XM_006252276.2 | XP_006252338.1 | 7 | 1372 | |||
| XM_006252277.2 | XP_006252339.1 | 7 | 969 | |||