siRNA Details
Gene Symbol: R3hcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252286.2 | XP_006252348.1 | 4 | 871 | ||
XM_006252288.2 | XP_006252350.1 | 4 | 874 | |||
XM_006252289.1 | XP_006252351.1 | 4 | 660 |
