siRNA Details
| Gene Symbol: Scel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108388.1 | NP_001101858.1 | 16 | 1065 | ||
| XM_006252428.2 | XP_006252490.1 | 16 | 1087 | |||
| XM_008770882.1 | XP_008769104.1 | 16 | 1069 | |||
| XM_008770883.1 | XP_008769105.1 | 16 | 1087 | |||

Scel
Genomic Map