siRNA Details
| Gene Symbol: Stk24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127494.1 | NP_001120966.1 | 281 | |||
| GenBank mRNA | EU371958.1 | - | 281 | |||
| Gene Symbol: LOC501266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759826.1 | XP_008758048.1 | 1 | 593 | ||

Stk24
Genomic Map