siRNA Details
| Gene Symbol: Zic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108392.2 | NP_001101862.2 | 4 | 2729 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009574, AK158319, AK161979, AK135125.

Zic2
Genomic Map